Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wnt5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022631.1 | NP_072153.1 | 2, 3 | 382 | ||
GenBank mRNA | AF188333.1 | - | 2, 3 | 382 |