Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nat8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022635.1 | NP_072157.1 | 1 | 641 | ||
XM_006236789.2 | XP_006236851.1 | 2 | 723 | |||
XM_006236790.2 | XP_006236852.1 | 2 | 733 | |||
GenBank mRNA | AF185570.1 | - | 1 | 641 |