Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crhr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022714.1 | NP_073205.1 | 3 | 508 | ||
XM_006236556.2 | XP_006236618.1 | 4 | 498 | |||
XM_006236557.2 | XP_006236619.1 | 4 | 498 | |||
XM_006236558.1 | XP_006236620.1 | 3 | 529 | |||
GenBank mRNA | EF078964.1 | - | 3 | 293 | ||
EF078965.1 | - | 3 | 293 | |||
U16253.1 | - | 3 | 508 |
Set Membership: |
![]() |