Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ireb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022863.2 | NP_074054.2 | 18 | 2352 | ||
XM_006243091.2 | XP_006243153.1 | 18 | 2657 | |||
XM_006243093.2 | XP_006243155.1 | 18 | 2332 | |||
GenBank mRNA | BC081798.1 | - | 18 | 2352 | 7130997 | |
U20181.1 | - | 18 | 2257 |