Gene Symbol: Map1lc3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022867.2 | NP_074058.2 | 2, 3 | 179 | ||
GenBank mRNA | AY206669.1 | - | 2, 3 | 141 | ||
AY392036.1 | - | 2, 3 | 179 | |||
BC058144.1 | - | 2, 3 | 137 | 6889616 | ||
BC083556.1 | - | 2, 3 | 159 | 7189855 | ||
FQ221245.1 | - | 2, 3 | 190 | |||
FQ228912.1 | - | 2, 3 | 190 | |||
U05784.1 | - | 2, 3 | 129 |