Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRPK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278273.1 | NP_001265202.1 | 15 | 1926 | ||
NM_182691.2 | NP_872633.1 | 13 | 1979 | |||
NM_182692.2 | NP_872634.1 | 14 | 1902 | |||
XM_005250549.3 | XP_005250606.2 | 15 | 2161 | |||
XM_005250550.3 | XP_005250607.1 | 14 | 1906 | |||
XM_005250551.4 | XP_005250608.1 | 15 | 1954 | |||
XM_006716098.2 | XP_006716161.1 | 15 | 1921 | |||
XM_011516536.2 | XP_011514838.1 | 13 | 1917 | |||
XM_011516538.2 | XP_011514840.1 | 14 | 1790 | |||
XM_017012560.1 | XP_016868049.1 | 15 | 1976 | |||
XM_017012561.1 | XP_016868050.1 | 14 | 1826 | |||
XM_017012562.1 | XP_016868051.1 | 15 | 2160 | |||
XM_017012563.1 | XP_016868052.1 | 15 | 2161 | |||
XM_017012564.1 | XP_016868053.1 | 13 | 1965 | |||
XM_017012566.1 | XP_016868055.1 | 15 | 1962 | |||
GenBank mRNA | AK093420.1 | - | 5 | 1701 | ||
AY354201.1 | - | 14 | 2068 | |||
BC035214.1 | - | 13 | 1898 | 4778251 | ||
BC068547.1 | - | 15 | 1926 | 5265599 | ||
U88666.1 | - | 13 | 1929 |