Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031730.2 | NP_113918.2 | 2 | 1700 | ||
XM_008762830.1 | XP_008761052.1 | 2 | 224 | |||
GenBank mRNA | CO393575.1 | - | 2 | 261 | ||
M59980.1 | - | 2 | 1379 | |||
S64320.1 | - | 2 | 1698 |