Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dync2h1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023024.1 | NP_075413.1 | 18 | 2815 | ||
GenBank mRNA | AB041881.1 | - | 18 | 2815 |