Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Arhgef9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023957.1 | NP_076447.1 | 8 | 1478 | ||
GenBank mRNA | AJ250425.1 | - | 8 | 1478 | ||
AJ302676.1 | - | 7 | 1298 |
Gene Symbol: LOC100912165 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773295.1 | XP_008771517.1 | 1 | 214 | ||
XM_008773296.1 | XP_008771518.1 | 217 |