Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gfra4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023967.1 | NP_076457.1 | 4 | 696 | ||
XM_006235040.2 | XP_006235102.1 | 3 | 2169 | |||
XM_006235041.2 | XP_006235103.1 | 3 | 2169 | |||
XM_006235042.2 | XP_006235104.1 | 3 | 2169 | |||
XM_006235043.2 | XP_006235105.1 | 3 | 2169 | |||
XM_006235045.2 | XP_006235107.1 | 4 | 1609 | |||
XM_008762196.1 | XP_008760418.1 | 4 | 1609 | |||
XM_008762197.1 | XP_008760419.1 | 4 | 1609 | |||
GenBank mRNA | AJ294475.1 | - | 4 | 696 | ||
AJ294476.1 | - | 4 | 696 |