Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Senp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023989.1 | NP_076479.1 | 16 | 1707 | ||
XM_008768805.1 | XP_008767027.1 | 16 | 1853 | |||
GenBank mRNA | AF260129.1 | - | 16 | 1707 | ||
FQ210639.1 | - | 16 | 1802 | |||
FQ212134.1 | - | 16 | 1801 | |||
FQ222500.1 | - | 15 | 1650 |