Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSTR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001049.2 | NP_001040.1 | 3 | 1420 | ||
GenBank mRNA | BC035618.1 | - | 1 | 1077 | 5175220 |
Set Membership: |
![]() |