Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tas2r13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023998.1 | NP_076488.1 | 1 | 133 | ||
GenBank mRNA | AF227145.1 | - | 1 | 133 |
Gene Symbol: Tas2r102 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006237482.1 | XP_006237544.1 | 1 | 193 |