Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Duox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024141.1 | NP_077055.1 | 10 | 1219 | ||
XM_006234862.2 | XP_006234924.1 | 12 | 2183 | |||
GenBank mRNA | AF237962.1 | - | 10 | 1219 |