Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSX2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164417.2 | NP_001157889.1 | 8 | 928 | ||
NM_001278701.1 | NP_001265630.1 | 9 | 1074 | |||
NM_001278702.1 | NP_001265631.1 | 9 | 1006 |
Gene Symbol: SSX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278697.1 | NP_001265626.1 | 9 | 1007 | ||
NM_003147.5 | NP_003138.3 | 9 | 1075 | |||
NM_175698.2 | NP_783629.1 | 8 | 929 | |||
GenBank mRNA | BC002818.2 | - | 9 | 1009 | 3636268 | |
BC007343.2 | - | 8 | 863 | 3615133 | ||
BC016957.1 | - | 8 | 863 | 3916128 | ||
BC071827.1 | - | 9 | 1009 | 4641362 | ||
Z49105.1 | - | 8 | 891 |