Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024373.1 | NP_077349.1 | 7, 8 | 1270 | ||
XM_006254471.2 | XP_006254533.1 | 8, 9 | 2691 | |||
GenBank mRNA | U35890.1 | - | 7, 8 | 1270 |