Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SS18 SIRNA Assays ›
Gene Symbol:
SS18
Gene Name:
SS18, nBAF chromatin remodeling complex subunit
Gene Aliases:
SSXT, SYT
Chromosome Location:
Chr.18: 26016253 - 26091217 on Build GRCh38
UniGene ID:
Hs.129261
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s13512
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SS18
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001007559.2
NP_001007560.1
4, 5
472
NM_001308201.1
NP_001295130.1
4, 5
447
NM_005637.3
NP_005628.2
4, 5
472
XM_006722527.2
XP_006722590.1
5, 6
498
XM_011526145.1
XP_011524447.1
4, 5
440
XM_011526147.2
XP_011524449.1
6, 7
723
XM_011526148.2
XP_011524450.1
6, 7
710
XM_011526149.2
XP_011524451.1
6, 7
735
XM_011526150.2
XP_011524452.1
5, 6
595
XM_011526151.2
XP_011524453.1
6, 7
724
XM_011526152.2
XP_011524454.1
5, 6
595
GenBank mRNA
AF244972.1
-
4, 5
409
AF343880.1
-
4, 5
406
AK289510.1
-
4, 5
450
AK296949.1
-
4, 5
430
AK299082.1
-
3, 4
340
AK301504.1
-
4, 5
477
AK304305.1
-
5, 6
588
AK307923.1
-
3, 4
363
AK316302.1
-
4, 5
422
AK316338.1
-
5, 6
541
AU279615.1
-
4, 5
477
BC096221.1
-
4, 5
409
40004908
BC096222.1
-
4, 5
409
40004909
BC096223.1
-
4, 5
409
40004911
BC096224.1
-
4, 5
409
40004912
CR542088.1
-
4, 5
406
CR542103.1
-
4, 5
406
DC378335.1
-
4, 5
447
X79201.1
-
4, 5
409
Back To Top
More Information
Panther Classification:
Molecular Function -
transcription cofactor
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
cell morphogenesis
transcription, DNA-templated
intracellular signal transduction
response to drug
positive regulation of transcription from RNA polymerase II promoter
ephrin receptor signaling pathway
neuronal stem cell population maintenance
protein binding
ligand-dependent nuclear receptor transcription coactivator activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-574f4745d9-9xb8z:80/10.239.165.207:80.
git-commit: d20c60ba4e48417b3d41b4fa5195ecf3ea66aacf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.12.0-2024.07.30-1.0