Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003752180.2 | XP_003752228.2 | 1, 2 | 2378 | ||
GenBank mRNA | L03556.1 | - | 1, 2 | 499 |
Gene Symbol: LOC103689922 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749749.2 | XP_003749797.2 | 2378 |
Gene Symbol: LOC103692127 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762946.1 | XP_008761168.1 | 1, 2 | 2378 |