Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ST5 SIRNA Assays ›
Gene Symbol:
ST5
Gene Name:
suppression of tumorigenicity 5
Gene Aliases:
DENND2B, HTS1, p126
Chromosome Location:
Chr.11: 8693352 - 8910951 on Build GRCh38
UniGene ID:
Hs.117715
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s13515
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ST5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_005418.3
NP_005409.3
20
3347
NM_139157.2
NP_631896.1
16
1815
NM_213618.1
NP_998783.1
17
3147
XM_005253077.1
XP_005253134.1
17
3062
XM_005253083.2
XP_005253140.1
16
1957
XM_011520309.1
XP_011518611.1
18
3122
XM_011520310.1
XP_011518612.1
18
3534
XM_011520311.2
XP_011518613.1
18
3516
XM_011520312.1
XP_011518614.1
22
3481
XM_011520313.1
XP_011518615.1
21
3367
XM_011520314.1
XP_011518616.1
20
3259
XM_011520315.1
XP_011518617.1
20
3355
XM_011520316.1
XP_011518618.1
19
3285
XM_011520317.1
XP_011518619.1
18
3226
XM_011520318.1
XP_011518620.1
21
3421
XM_011520319.2
XP_011518621.1
18
3074
XM_011520323.2
XP_011518625.2
17
1945
XM_011520324.2
XP_011518626.2
16
1885
XM_011520325.1
XP_011518627.1
17
2017
XM_011520326.1
XP_011518628.1
21
2221
XM_011520327.1
XP_011518629.1
17
1910
XM_011520328.1
XP_011518630.1
17
1942
XM_011520329.1
XP_011518631.1
20
2161
XM_017018182.1
XP_016873671.1
19
3240
XM_017018183.1
XP_016873672.1
19
3217
XM_017018184.1
XP_016873673.1
21
3376
XM_017018185.1
XP_016873674.1
17
3172
XM_017018186.1
XP_016873675.1
16
3112
XM_017018187.1
XP_016873676.1
17
1881
GenBank mRNA
AK127763.1
-
14
2202
AK293245.1
-
14
1580
AK307617.1
-
6
702
AK312305.1
-
17
3113
AK312758.1
-
16
2157
BC036655.1
-
20
3352
4837231
BX647431.1
-
17
3147
S45936.1
-
13
1485
U15131.1
-
17
3075
U15779.1
-
13
1444
U15780.1
-
16
1815
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of GTPase activity
Rab guanyl-nucleotide exchange factor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-qqtbl:80/10.239.164.61:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0