Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2d4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031001.2 | NP_112263.1 | 1 | 339 | ||
GenBank mRNA | BC078808.1 | - | 1 | 339 | 7113229 | |
U56407.1 | - | 1 | 230 |