Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053331.2 | NP_445783.1 | 2 | 282 | ||
GenBank mRNA | BC081760.1 | - | 2 | 282 | 7190504 | |
FQ214521.1 | - | 2 | 308 | |||
FQ214678.1 | - | 2 | 308 | |||
FQ214699.1 | - | 2 | 288 | |||
FQ214706.1 | - | 2 | 320 | |||
FQ214836.1 | - | 2 | 308 | |||
FQ215326.1 | - | 2 | 246 | |||
FQ215775.1 | - | 2 | 317 | |||
FQ216090.1 | - | 2 | 288 | |||
FQ216404.1 | - | 2 | 286 | |||
FQ217268.1 | - | 2 | 286 | |||
FQ220081.1 | - | 2 | 320 | |||
FQ223556.1 | - | 2 | 295 | |||
FQ229711.1 | - | 2 | 246 | |||
U73525.1 | - | 2 | 276 |