Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cyb5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030586.2 | NP_085075.1 | 3, 4 | 319 | ||
GenBank mRNA | BC072535.1 | - | 3, 4 | 319 | 7103272 | |
X96392.1 | - | 3, 4 | 187 | |||
Y12517.1 | - | 3, 4 | 364 |