Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037295.1 | NP_001032372.1 | 2 | 1114 | ||
NM_030831.1 | NP_110458.1 | 1 | 1284 | |||
GenBank mRNA | BC127447.1 | - | 2 | 1042 | 8363428 | |
U12184.1 | - | 1 | 1284 | |||
X61496.1 | - | 2 | 1111 |
Set Membership: |
![]() |