Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031008.2 | NP_112270.2 | 4 | 407 | ||
XM_008760010.1 | XP_008758232.1 | 4 | 465 | |||
GenBank mRNA | BC081786.1 | - | 4 | 407 | 7126468 | |
X53773.1 | - | 363 |