Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031011.3 | NP_112273.3 | 7 | 1179 | ||
GenBank mRNA | AY724509.1 | - | 7 | 862 | ||
BC061532.1 | - | 7 | 1182 | 6921099 | ||
FM123317.1 | - | 2 | 376 | |||
FQ232080.1 | - | 7 | 999 | |||
FQ234346.1 | - | 7 | 1002 | |||
FQ234398.1 | - | 2 | 1348 | |||
M34464.1 | - | 7 | 1125 |