Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabbr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031028.3 | NP_112290.2 | 17 | 2249 | ||
XM_006255879.2 | XP_006255941.1 | 17 | 2343 | |||
XM_008772700.1 | XP_008770922.1 | 16 | 2322 | |||
GenBank mRNA | AB016160.1 | - | 12 | 1792 | ||
AB016161.1 | - | 12 | 1792 | |||
AF283276.1 | - | 16 | 2163 | |||
AJ831471.1 | - | 17 | 2247 | |||
AJ831472.1 | - | 17 | 2371 | |||
Y10369.2 | - | 17 | 2221 | |||
Y10370.2 | - | 12 | 1894 |
Set Membership: |
G-protein coupled receptor |