Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Gmfb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031032.2 | NP_112294.1 | 6 | 429 | ||
XM_006251814.2 | XP_006251876.1 | 6 | 1093 | |||
GenBank mRNA | BC081778.1 | - | 6 | 429 | 7192655 | |
FQ222145.1 | - | 6 | 491 |