Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: STC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003155.2 | NP_003146.1 | 4 | 828 | ||
GenBank mRNA | AK297734.1 | - | 5 | 675 | ||
AK309366.1 | - | 4 | 692 | |||
AK312589.1 | - | 4 | 827 | |||
BC029044.1 | - | 4 | 806 | 5191420 | ||
KP455655.1 | - | 4 | 491 | |||
KP455656.1 | - | 4 | 491 | |||
KP455657.1 | - | 4 | 491 | |||
KP455658.1 | - | 4 | 492 | |||
KP455659.1 | - | 4 | 491 | |||
U25997.1 | - | 4 | 828 | |||
U46768.1 | - | 4 | 578 |