Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pros1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031086.2 | NP_112348.2 | 7 | 798 | ||
XM_006240719.2 | XP_006240781.1 | 5 | 1084 | |||
XM_008765045.1 | XP_008763267.1 | 5 | 530 | |||
GenBank mRNA | CB580854.1 | - | 2 | 155 | ||
DY571157.1 | - | 6 | 619 |