Gene Symbol: Stx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031124.1 | NP_112386.1 | 8, 9 | 754 | ||
XM_008760263.1 | XP_008758485.1 | 8, 9 | 838 | |||
XM_008760264.1 | XP_008758486.1 | 8, 9 | 838 | |||
XM_008760265.1 | XP_008758487.1 | 8, 9 | 838 | |||
XM_008760267.1 | XP_008758489.1 | 8, 9 | 838 | |||
GenBank mRNA | BC168246.1 | - | 8, 9 | 708 | 5600098 | |
L20820.1 | - | 8, 9 | 754 |