Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyr61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031327.2 | NP_112617.2 | 4 | 930 | ||
GenBank mRNA | AB015877.1 | - | 4 | 795 | ||
AF218568.1 | - | 4 | 930 | |||
BC081689.1 | - | 4 | 910 | 7122117 |