Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031328.1 | NP_112618.1 | 2 | 225 | ||
GenBank mRNA | AB016069.1 | - | 2 | 225 | ||
BC061772.1 | - | 2 | 213 | 5621919 |