Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031331.1 | NP_112621.1 | 5 | 483 | ||
XM_006232924.2 | XP_006232986.1 | 5 | 488 | |||
GenBank mRNA | AB017188.1 | - | 5 | 483 | ||
AF175575.1 | - | 5 | 424 | |||
BC060559.1 | - | 5 | 433 | 6891370 | ||
FQ209672.1 | - | 4 | 285 |