Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mkln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031359.1 | NP_112649.1 | 1, 2 | 103 | ||
GenBank mRNA | AB046442.1 | - | 1, 2 | 103 |