Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pax4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031799.1 | NP_113987.1 | 8 | 781 | ||
XM_006236231.2 | XP_006236293.1 | 9 | 6834 | |||
XM_006236232.2 | XP_006236294.1 | 10 | 1830 | |||
XM_006236233.2 | XP_006236295.1 | 10 | 1272 | |||
GenBank mRNA | AF053100.1 | - | 8 | 781 | ||
AF053101.1 | - | 7 | 655 | |||
AF198155.1 | - | 7 | 733 |