Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mybph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031813.1 | NP_114001.1 | 4 | 578 | ||
XM_006249888.2 | XP_006249950.1 | 4 | 688 | |||
XM_006249889.2 | XP_006249951.1 | 4 | 687 | |||
GenBank mRNA | AF077338.1 | - | 4 | 578 | ||
BC061993.1 | - | 4 | 617 | 5599482 |