Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Git1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031814.1 | NP_114002.1 | 11 | 1083 | ||
GenBank mRNA | AF085693.1 | - | 11 | 1083 |