Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rbbp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031816.1 | NP_114004.1 | 9 | 1277 | ||
GenBank mRNA | AF090306.1 | - | 9 | 1277 | ||
BC062012.1 | - | 9 | 1186 | 5621292 |