Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctcf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031824.1 | NP_114012.1 | 6 | 1283 | ||
XM_006255536.1 | XP_006255598.1 | 5 | 1233 | |||
XM_008772536.1 | XP_008770758.1 | 4 | 1250 | |||
XM_008772537.1 | XP_008770759.1 | 4 | 1250 | |||
XM_008772538.1 | XP_008770760.1 | 4 | 1250 | |||
XM_008772539.1 | XP_008770761.1 | 4 | 1250 | |||
GenBank mRNA | AF133731.2 | - | 6 | 1319 |