Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fetub | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053348.1 | NP_445800.1 | 4 | 637 | ||
XM_006248549.1 | XP_006248611.1 | 5 | 591 | |||
XM_006248550.1 | XP_006248612.1 | 5 | 954 | |||
GenBank mRNA | AJ242926.1 | - | 4 | 637 | ||
BC070508.1 | - | 5 | 576 | 7099154 | ||
FQ209398.1 | - | 4 | 648 | |||
FQ209449.1 | - | 5 | 572 | |||
FQ209518.1 | - | 4 | 588 | |||
FQ209610.1 | - | 4 | 623 | |||
FQ209832.1 | - | 4 | 588 | |||
FQ210294.1 | - | 4 | 588 | |||
FQ210335.1 | - | 4 | 589 | |||
FQ211050.1 | - | 4 | 588 | |||
FQ211248.1 | - | 4 | 623 | |||
FQ218476.1 | - | 4 | 619 | |||
FQ218557.1 | - | 5 | 567 | |||
FQ218709.1 | - | 4 | 588 | |||
FQ218884.1 | - | 5 | 559 | |||
FQ218893.1 | - | 4 | 588 | |||
FQ218930.1 | - | 4 | 621 | |||
FQ218949.1 | - | 4 | 438 | |||
FQ219048.1 | - | 4 | 619 | |||
FQ219152.1 | - | 4 | 625 | |||
FQ219170.1 | - | 4 | 588 | |||
FQ219190.1 | - | 4 | 623 | |||
FQ219266.1 | - | 4 | 588 | |||
FQ219426.1 | - | 2 | 190 | |||
FQ219454.1 | - | 4 | 588 | |||
FQ219459.1 | - | 4 | 586 | |||
FQ219500.1 | - | 4 | 591 | |||
FQ219513.1 | - | 4 | 660 |