Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pon1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032077.1 | NP_114466.1 | 6 | 625 | ||
GenBank mRNA | BC091403.1 | - | 6 | 666 | 7324332 | |
FQ209439.1 | - | 6 | 643 | |||
FQ209632.1 | - | 3 | 321 | |||
FQ211027.1 | - | 6 | 636 | |||
FQ211041.1 | - | 6 | 679 | |||
FQ218360.1 | - | 6 | 641 | |||
FQ218888.1 | - | 6 | 636 | |||
FQ219478.1 | - | 6 | 628 | |||
FQ219506.1 | - | 6 | 636 | |||
FQ219654.1 | - | 6 | 628 | |||
FQ219750.1 | - | 6 | 630 | |||
FQ219776.1 | - | 6 | 636 | |||
FQ223824.1 | - | 5 | 536 | |||
U94856.1 | - | 6 | 583 |