Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ssbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053358.1 | NP_445810.1 | 10, 11 | 1027 | ||
XM_006238534.2 | XP_006238596.1 | 11, 12 | 1166 | |||
XM_006238535.2 | XP_006238597.1 | 11, 12 | 1140 | |||
XM_006238538.2 | XP_006238600.1 | 10, 11 | 1082 | |||
XM_008763905.1 | XP_008762127.1 | 11, 12 | 2817 | |||
XM_008763906.1 | XP_008762128.1 | 11, 12 | 2790 | |||
XM_008763909.1 | XP_008762131.1 | 10, 11 | 2706 | |||
GenBank mRNA | AF121893.1 | - | 10, 11 | 1027 | ||
AY724490.1 | - | 11, 12 | 808 | |||
BC107666.1 | - | 10, 11 | 1011 | 7936459 | ||
FQ230168.1 | - | 5, 6 | 305 |