Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SURF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278928.1 | NP_001265857.1 | 5, 6 | 744 | ||
NM_017503.4 | NP_059973.4 | 6 | 747 | |||
GenBank mRNA | BC014411.1 | - | 6 | 716 | 4548425 | |
BQ672017.1 | - | 4 | 394 | |||
CR456753.1 | - | 6 | 699 | |||
Z35094.1 | - | 6 | 716 |