Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clcn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053363.2 | NP_445815.2 | 7 | 962 | ||
XM_006253070.2 | XP_006253132.1 | 7 | 1118 | |||
XM_006253071.2 | XP_006253133.1 | 6 | 1066 | |||
XM_006253073.2 | XP_006253135.1 | 6 | 4042 | |||
XM_006253075.2 | XP_006253137.1 | 6 | 1066 | |||
XM_006253076.2 | XP_006253138.1 | 6 | 1958 | |||
XM_006253077.2 | XP_006253139.1 | 6 | 1064 | |||
GenBank mRNA | AF142778.1 | - | 7 | 962 | ||
D17521.1 | - | 6 | 1081 |