Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other IHH SIRNA Assays ›
Gene Symbol:
Ihh
Gene Name:
Indian hedgehog
Gene Aliases:
-
Chromosome Location:
Chr.9: 82208223 - 82214440 on Build Rnor_6.0
UniGene ID:
Rn.8711
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s136542
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Ihh
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_053384.1
NP_445836.1
3
1081
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_010544, U85610, BC046984, AK144322, X76291.
Gene Ontology Categories:
Function(s)
Process(es)
skeletal system development
ossification
patterning of blood vessels
osteoblast differentiation
in utero embryonic development
cell fate specification
morphogenesis of a branching structure
vasculature development
heart looping
positive regulation of mesenchymal cell proliferation
epithelial cell morphogenesis
retinal pigment epithelium development
proteoglycan metabolic process
proteolysis
smoothened signaling pathway
cell-cell signaling
pattern specification process
positive regulation of cell proliferation
embryonic pattern specification
negative regulation of signal transduction
intein-mediated protein splicing
cell differentiation
vitelline membrane formation
pancreas development
positive regulation of chondrocyte differentiation
response to estradiol
positive regulation of collagen biosynthetic process
negative regulation of T cell differentiation in thymus
negative regulation of immature T cell proliferation in thymus
positive regulation of T cell differentiation in thymus
multicellular organism growth
chondrocyte proliferation
regulation of growth
embryonic digit morphogenesis
camera-type eye development
negative regulation of apoptotic process
bone resorption
regulation of cell differentiation
negative regulation of cell differentiation
positive regulation of smoothened signaling pathway
positive regulation of transcription from RNA polymerase II promoter
positive regulation of alpha-beta T cell differentiation
negative regulation of alpha-beta T cell differentiation
negative regulation of eye pigmentation
cell maturation
embryonic digestive tract morphogenesis
embryonic camera-type eye morphogenesis
neuron development
smooth muscle tissue development
positive regulation of epithelial cell proliferation
cartilage development
maternal process involved in female pregnancy
camera-type eye photoreceptor cell fate commitment
head morphogenesis
somite development
embryonic skeletal joint development
epithelial cell-cell adhesion
molecular_function
patched binding
calcium ion binding
peptidase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-qtpg5:80/10.239.161.70:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline