Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdh8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053393.2 | NP_445845.2 | 2 | 643 | ||
GenBank mRNA | AB010436.1 | - | 2 | 643 | ||
AB010437.1 | - | 2 | 643 |