Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adcy7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053396.1 | NP_445848.1 | 20 | 2648 | ||
XM_008772421.1 | XP_008770643.1 | 20 | 2665 |