Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Strbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053416.2 | NP_445868.2 | 8, 9 | 1109 | ||
XM_006234108.2 | XP_006234170.2 | 9, 10 | 1447 | |||
XM_008761777.1 | XP_008759999.1 | 9, 10 | 1147 | |||
XM_008761778.1 | XP_008760000.1 | 9, 10 | 1147 | |||
GenBank mRNA | BC078774.1 | - | 7, 8 | 731 | ||
CA512540.1 | - | 5, 6 | 538 |