Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc4a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053424.1 | NP_445876.1 | 14, 15 | 1964 | ||
XM_006250788.2 | XP_006250850.1 | 14, 15 | 2424 | |||
XM_006250789.2 | XP_006250851.1 | 14, 15 | 2424 | |||
XM_006250790.2 | XP_006250852.1 | 15, 16 | 2142 | |||
XM_006250791.2 | XP_006250853.1 | 14, 15 | 1976 | |||
GenBank mRNA | AF004017.1 | - | 11, 12 | 1802 | ||
AF124441.1 | - | 14, 15 | 1964 | |||
AF210250.1 | - | 14, 15 | 1971 | |||
AF254802.1 | - | 14, 15 | 1964 |