Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sf3b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053426.1 | NP_445878.1 | 16 | 2381 | ||
XM_006244933.2 | XP_006244995.1 | 16 | 2378 | |||
XM_006244934.2 | XP_006244996.1 | 13 | 3286 |